Learn More
Copper(II) chloride dihydrate, 99%, Thermo Scientific Chemicals
$127.60 - $320.38
Chemical Identifiers
CAS | 10125-13-0 |
---|---|
Molecular Formula | Cl2CuH4O2 |
Molecular Weight (g/mol) | 170.48 |
MDL Number | MFCD00149674 |
InChI Key | MPTQRFCYZCXJFQ-UHFFFAOYSA-L |
Synonym | coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 |
PubChem CID | 61482 |
ChEBI | CHEBI:86318 |
IUPAC Name | dichlorocopper;dihydrate |
SMILES | O.O.[Cl-].[Cl-].[Cu++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1606430
|
Thermo Scientific Chemicals
A1606430 |
250 g |
Each for $127.60
|
|
|||||
AAA160640B
|
Thermo Scientific Chemicals
A160640B |
1000 g |
Each for $320.38
|
|
|||||
Description
Copper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCopper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.
Solubility
Soluble in water, methanol and ethanol.
Notes
Incompatible with materials such as oxidizing agents, metals and acids.
Chemical Identifiers
10125-13-0 | |
170.48 | |
MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
61482 | |
dichlorocopper;dihydrate |
Cl2CuH4O2 | |
MFCD00149674 | |
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 | |
CHEBI:86318 | |
O.O.[Cl-].[Cl-].[Cu++] |
Specifications
100°C (decomposition) | |
3.0 to 3.8 | |
Cl2CuH4O2 | |
UN2802 | |
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 | |
MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
dichlorocopper;dihydrate | |
61482 | |
170.48 (134.45 Anhydrous) | |
Odorless | |
2.51 |
10125-13-0 | |
250 g | |
MFCD00149674 | |
14,2633 | |
Soluble in water,methanol and ethanol. | |
O.O.[Cl-].[Cl-].[Cu++] | |
170.48 | |
CHEBI:86318 | |
99% | |
Hygroscopic | |
Copper(II) chloride dihydrate |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H318
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: COPPER CHLORIDE
EINECSNumber : 231-210-2
RTECSNumber : GL7030000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only