missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) ethylacetoacetate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04148918

 View more versions of this product

Catalog No. AA4148918


Explore available promotions

Description

Description

Copper(II) ethylacetoacetate it acts as a common cu(+2) precursor. It is also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) ethylacetoacetate
192°C
C12H18CuO6
50 g
OXFZSJOUPRCPJO-UHFFFAOYSA-N
321.81
Powder
14284-06-1
Odorless
MFCD00017759
Soluble in ethanol
321.82
97%

Chemical Identifiers

14284-06-1
321.82
OXFZSJOUPRCPJO-UHFFFAOYSA-N
C12H18CuO6
MFCD00017759
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 238-181-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only