missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) ethylacetoacetate, 97%, Thermo Scientific Chemicals

$58.12 - $835.41

Chemical Identifiers

CAS 14284-06-1
Molecular Formula C12H18CuO6
Molecular Weight (g/mol) 321.82
MDL Number MFCD00017759
InChI Key OXFZSJOUPRCPJO-UHFFFAOYSA-N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4148904
View Documents
Thermo Scientific Chemicals
04148904
2 g
Each for $58.12
 
AA4148909
View Documents
Thermo Scientific Chemicals
04148909
10 g
Each for $206.10
 
AA4148918
View Documents
Thermo Scientific Chemicals
04148918
50 g
Each for $835.41
 
Description

Description

Copper(II) ethylacetoacetate it acts as a common cu(+2) precursor. It is also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14284-06-1
321.82
OXFZSJOUPRCPJO-UHFFFAOYSA-N
C12H18CuO6
MFCD00017759

Specifications

14284-06-1
Odorless
MFCD00017759
Soluble in ethanol
321.82
97%
Copper(II) ethylacetoacetate
192°C
C12H18CuO6
2 g
OXFZSJOUPRCPJO-UHFFFAOYSA-N
321.81
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 238-181-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only