missing translation for 'onlineSavingsMsg'
Learn More

Copper(II)-ethylenediamine complex, 1M solution in water

CAS: 14552-35-3 | C4H18CuN4O2 | 217.76 g/mol

Supplier:  Thermo Scientific Chemicals 258550010

 View more versions of this product

Catalog No. AC258550010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II)-ethylenediamine complex
14552-35-3,7732-18-5
C4H18CuN4O2
MFCD00274628
Bis(ethylenediamine)copper(II)hydroxide
217.76
Glass bottle
1.1000g/mL
1M Solution in Water
1 L
Cu(H2NCH2CH2NH2)2(OH)2
>110°C
Solubility in water: soluble
217.76
1.1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only