missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(II)-ethylenediamine complex, 1M solution in water, Thermo Scientific Chemicals
$95.62 - $95.62
Specifications
Chemical Name or Material | Copper(II)-ethylenediamine complex |
---|---|
Name Note | 1M Solution in Water |
CAS | 14552-35-3,7732-18-5 |
Molecular Formula | C4H18CuN4O2 |
Linear Formula | Cu(H2NCH2CH2NH2)2(OH)2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC258550010
|
Thermo Scientific Chemicals
258550010 |
1 L | Glass bottle |
Each for $95.62
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II)-ethylenediamine complex | |
14552-35-3,7732-18-5 | |
Cu(H2NCH2CH2NH2)2(OH)2 | |
>110°C | |
Solubility in water: soluble | |
217.76 | |
1.1 |
1M Solution in Water | |
C4H18CuN4O2 | |
MFCD00274628 | |
Bis(ethylenediamine)copper(II)hydroxide | |
217.76 | |
Glass bottle | |
1.1000g/mL |
RUO – Research Use Only