missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) isopropoxide, 98% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03582106

 View more versions of this product

Catalog No. AA3582106

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) isopropoxide
(metals basis)
MFCD00074982
UN3180
copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
VNGORJHUDAPOQZ-UHFFFAOYSA-N
copper;propan-2-olate
14389592
98%
23578-23-6
C6H14CuO2
5 g
Moisture sensitive
Hydrolyses in water.
[Cu++].CC(C)[O-].CC(C)[O-]
181.72
181.73
Powder

Chemical Identifiers

23578-23-6
181.72
VNGORJHUDAPOQZ-UHFFFAOYSA-N
14389592
[Cu++].CC(C)[O-].CC(C)[O-]
C6H14CuO2
MFCD00074982
copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
copper;propan-2-olate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302-H315-H319-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only