missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) isopropoxide, 98% (metals basis), Thermo Scientific Chemicals

$401.75 - $1647.22

Chemical Identifiers

CAS 23578-23-6
Molecular Formula C6H14CuO2
Molecular Weight (g/mol) 181.72
MDL Number MFCD00074982
InChI Key VNGORJHUDAPOQZ-UHFFFAOYSA-N
Synonym copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
PubChem CID 14389592
IUPAC Name copper;propan-2-olate
SMILES [Cu++].CC(C)[O-].CC(C)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3582106
View Documents
Thermo Scientific Chemicals
03582106
5 g
Each for $401.75
 
AA3582114
View Documents
Thermo Scientific Chemicals
03582114
25 g
Each for $1,647.22
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23578-23-6
181.72
VNGORJHUDAPOQZ-UHFFFAOYSA-N
14389592
[Cu++].CC(C)[O-].CC(C)[O-]
C6H14CuO2
MFCD00074982
copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
copper;propan-2-olate

Specifications

23578-23-6
MFCD00074982
UN3180
copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
VNGORJHUDAPOQZ-UHFFFAOYSA-N
copper;propan-2-olate
14389592
98%
Copper(II) isopropoxide
C6H14CuO2
5 g
Moisture sensitive
Hydrolyses in water.
[Cu++].CC(C)[O-].CC(C)[O-]
181.72
181.73
Powder
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302-H315-H319-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only