missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) phosphate, 98%

CAS: 7798-23-4 | Cu3O8P2 | 380.578 g/mol

$100.10 - $1,733.53

Chemical Identifiers

CAS 7798-23-4
Molecular Formula Cu3O8P2
Molecular Weight (g/mol) 380.578
MDL Number MFCD00036268
InChI Key GQDHEYWVLBJKBA-UHFFFAOYSA-H
Synonym copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
PubChem CID 86469
IUPAC Name tricopper;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1736022
View Documents
Thermo Scientific Chemicals
A1736022
100 g N/A
Only null left
 
AAA1736036
View Documents
Thermo Scientific Chemicals
A1736036
500 g N/A
Only null left
 
AAA173600E
View Documents
Thermo Scientific Chemicals
A173600E
2500 g N/A
Only null left
 
Description

Description

Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.

Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.
Specifications

Chemical Identifiers

7798-23-4
380.578
GQDHEYWVLBJKBA-UHFFFAOYSA-H
86469
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]
Cu3O8P2
MFCD00036268
copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
tricopper;diphosphate

Specifications

>300°C
100 g
Cu3(PO4)2
14,2648
Insoluble with water and ethanol. Soluble in ammonia,ammonium hydroxide,acetone.
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]
380.578
380.58
Odorless
7798-23-4
Cu3O8P2
MFCD00036268
copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
GQDHEYWVLBJKBA-UHFFFAOYSA-H
tricopper;diphosphate
86469
98%
Copper(II) phosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 232-254-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only