missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 042925A1

 View more versions of this product

Catalog No. AA42925A1


Explore available promotions

Description

Description

Copper(II) sulfide is used in antifouling paints, mixed catalysts and to develop aniline black dyes in textile printing. Further, it is used as a varnish pigment and an oxidizing agent. In addition, it is used as a component of solid state electrodes and thin-film solar cell with cadmium sulfide. It is a moderate conductor of electricity and used in photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) sulfide
1317-40-4
Powder
99.8% (Metal basis)
MFCD00016066
Soluble in nitric acid,ammonium hydroxide,potassium cyanide. Insoluble in water,ethanol,hydrochloric acid ,dilute acids,sulfuric acid,alkali cyanides,aqueous ammonia and alkaline solutions.
[S--].[Cu++]
95.61
95.6
1.45
Powder
220°C (decomposition)
Black
1 kg
CuS
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
OMZSGWSJDCOLKM-UHFFFAOYSA-N
copper(2+) sulfanediide
14831
Odorless
Air and moisture sensitive
3.9-4.6

Chemical Identifiers

1317-40-4
95.61
OMZSGWSJDCOLKM-UHFFFAOYSA-N
14831
CuS
MFCD00016066
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
[S--].[Cu++]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-271-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only