Learn More
Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
$153.03 - $1482.86
Chemical Identifiers
CAS | 1317-40-4 |
---|---|
Molecular Formula | CuS |
Molecular Weight (g/mol) | 95.61 |
MDL Number | MFCD00016066 |
InChI Key | OMZSGWSJDCOLKM-UHFFFAOYSA-N |
Synonym | copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue |
PubChem CID | 14831 |
SMILES | [S--].[Cu++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA4292518
|
Thermo Scientific Chemicals
04292518 |
50 g |
Each for $153.03
|
|
||||||
AA4292530
|
Thermo Scientific Chemicals
04292530 |
250 g |
Each for $452.89
|
|
||||||
AA42925A1
|
Thermo Scientific Chemicals
042925A1 |
1 kg |
Each for $1,482.86
|
|
||||||
Description
Copper(II) sulfide is used in antifouling paints, mixed catalysts and to develop aniline black dyes in textile printing. Further, it is used as a varnish pigment and an oxidizing agent. In addition, it is used as a component of solid state electrodes and thin-film solar cell with cadmium sulfide. It is a moderate conductor of electricity and used in photovoltaics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-40-4 | |
95.61 | |
OMZSGWSJDCOLKM-UHFFFAOYSA-N | |
14831 |
CuS | |
MFCD00016066 | |
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue | |
[S--].[Cu++] |
Specifications
220°C (decomposition) | |
Black | |
50 g | |
CuS | |
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue | |
OMZSGWSJDCOLKM-UHFFFAOYSA-N | |
copper(2+) sulfanediide | |
14831 | |
Odorless | |
Air and moisture sensitive | |
4.6 g/cm3 |
1317-40-4 | |
Powder | |
99.8% (Metal basis) | |
MFCD00016066 | |
Soluble in nitric acid,ammonium hydroxide,potassium cyanide. Insoluble in water,ethanol,hydrochloric acid ,dilute acids,sulfuric acid,alkali cyanides,aqueous ammonia and alkaline solutions. | |
[S--].[Cu++] | |
95.61 | |
95.6 | |
1.45 | |
Powder | |
Copper(II) sulfide |
Safety and Handling
EINECSNumber : 215-271-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only