missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Supplier: Corning™ C4511
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
16 x 24 | |
Black | |
384 | |
Non-sterile | |
Round Bottom (U-well) |
Without Lid | |
Medium Binding | |
Polystyrene | |
2 to 25 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. |