missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
$767.19 - $2532.06
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Color | Well Shape | Surface Treatment | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Well Shape | Surface Treatment | Price | Quantity | |||
09-761-21
|
Corning™
C3577 |
Black | Flat Bottom (F-well) | High Binding |
Case of 50 for $767.19
|
|
|||
07-201-322
|
Corning™
C3574 |
White | Flat Bottom (F-well) | Non-binding |
Case of 50 Each for $1,620.80
|
|
|||
09-761-86
|
Corning™
3575 |
Black | Flat Bottom (F-well) | Non-binding |
Case of 50 Each for $1,753.73
|
|
|||
09-761-85
|
Corning™
C3573 |
Black | Flat Bottom (F-well) | Non-treated |
Case of 50 Each for $1,268.95
|
|
|||
15-100-155
|
Corning™
C4510 |
Black | Round Bottom (U-well) | High Binding |
Case of 50 Each for $2,227.67
|
|
|||
15-100-157
|
Corning™
4512 |
White | Round Bottom (U-well) | Non-treated |
Case of 50 Each for $1,959.97
|
|
|||
15-100-156
|
Corning™
C4511 |
Black | Round Bottom (U-well) | Non-treated |
Case of 50 Each for $1,039.21
|
|
|||
15-100-159
|
Corning™
4514 |
Black | Round Bottom (U-well) | Non-binding |
Case of 50 Each for $2,340.21
|
|
|||
15-100-158
|
Corning™
4513 |
White | Round Bottom (U-well) | Non-binding |
Case of 50 Each for $2,532.06
|
|
|||
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
Polystyrene | |
Non-sterile |
16 x 24 | |
384 |