Corning™ 96-Well, Flat-Bottom Polystyrene NBS Microplate Without Lid
Nonsterile
Supplier: Corning™ 3991
Description
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Non-binding | |
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise | |
5/Pk. |
Without Lid | |
Black | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
25/Cs. | |
25/Cs. |