missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 96-Well, Flat-Bottom Polystyrene NBS Microplate Without Lid
Nonsterile
$851.08 - $917.00
Specifications
Lid | Without Lid |
---|---|
Array | 8 x 12 |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Color | Volume (Metric) Working | Format | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Volume (Metric) Working | Format | Price | Quantity | |||
07-201-203
|
Corning™
3991 |
Black | 75 to 200 μL | Standard |
Case of 25 Each for $851.08
|
|
|||
07-201-204
|
Corning™
3992 |
White | 25 to 125 μL | Half-Area |
Case of 25 Each for $917.00
|
|
|||
Description
Specifications
Without Lid | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) | |
25/Cs. | |
25/Cs. |
8 x 12 | |
96 | |
Non-binding | |
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise | |
5/Pk. |