Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Supplier: Corning™ 354551
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Collagen Type I | |
Lid | |
150 mm |
Closure Included | |
Round | |
151.9 sq. cm | |
Round | |
Cell Culture Dish | |
5/cs. |