Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
$218.20 - $904.59
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Round
Color Clear
View More Specs
Products 7
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity  
08-772-74
View Documents
Corning™
356401
60 mm 100/cs.
Case of 100 Each for $904.59
08-772-73
View Documents
Corning™
356456
35 mm 100/cs.
Case of 100 Each for $788.14
08-774-6
View Documents
Corning™
354456
35 mm 20/cs.
Case of 20 Each for $306.97
08-774-7
View Documents
Corning™
354401
60 mm 20/cs.
Case of 20 Each for $218.20
08-774-8
View Documents
Corning™
354450
100 mm 10/cs.
Case of 10 Each for $303.48
08-774-9
View Documents
Corning™
354551
150 mm 5/cs.
Case of 5 Each for $465.81
08-772-75
View Documents
Corning™
356450
100 mm 40/cs.
Case of 40 Each for $683.27
Description
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
Clear
Polystyrene
Clear
Round
Cell Culture Dish
Closure Included
Round
Polystyrene
Collagen Type I
Lid
SDS
missing translation for 'documents'
missing translation for 'documents'