Corning™ Black 384-Well Polypropylene Assay Plate
Lower compound sticking and higher solvent resistance than polystyrene plates
$2533.69 - $2533.69
Specifications
Format Standard
Array 16 x 24
Color Black
Material Polypropylene
No. of Wells 384
View More Specs
Products 1
Catalog Number Mfr. No. Lid Sterility Price Quantity  
Catalog Number Mfr. No. Lid Sterility Price Quantity  
07-200-765
View Documents
Corning™
3658
Without Lid Non-sterile
Case of 100 Each for $2,533.69
Description
Description
  • Ideal for fluorescence assays and for compound storage
  • Round-bottom wells
  • Untreated
Specifications
Specifications
Standard
Black
384
16 x 24
Polypropylene
For fluorescence assays and compound storage
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'