missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Sterile Cell Strainers
Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.
Supplier: Corning™ 431751
Description
- Strong nylon mesh available in three pore sizes (40, 70 or 100 μm) for variety of applications
- Fits all major brands of 50 mL conical tubes
- Non cytotoxic device has color-coded polypropylene frames
- Grip area for handling with forceps
- Sterilized by gamma irradiation
- Use for pre-filtering solutions containing particulates >40μm and for cleaning cell suspensions prior to sub culturing, counting and analysis or freezing
- Individually packed in easy to open bags
For preparation of:
- Single cell suspensions of blood cells
- Specimens for primary cell cultures and immunogens
- Primary cells from tissue
Specifications
Nylon | |
Polypropylene | |
Conical Tubes, 50 mL |
White | |
Sterile | |
70 μm |