missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Sterile Cell Strainers

Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.

$394.92 - $405.13

Specifications

Mesh Material Nylon
Material Polypropylene
Sterility Sterile
For Use With (Equipment) Conical Tubes, 50 mL
View More Specs

Products 3
Catalog Number Mfr. No. Color Mesh Size Price Quantity  
Catalog Number Mfr. No. Color Mesh Size Price Quantity  
07-201-430
View Documents
Corning™
431750
Blue 40 μm
Case of 50 Each for $401.96
 
07-201-431
View Documents
Corning™
431751
White 70 μm
Case of 50 Each for $394.92
 
07-201-432
View Documents
Corning™
431752
Yellow 100 μm
Case of 50 Each for $405.13
 
Description

Description

 

  • Strong nylon mesh available in three pore sizes (40, 70 or 100 μm) for variety of applications
  • Fits all major brands of 50 mL conical tubes
  • Non cytotoxic device has color-coded polypropylene frames
  • Grip area for handling with forceps
  • Sterilized by gamma irradiation
  • Use for pre-filtering solutions containing particulates >40μm and for cleaning cell suspensions prior to sub culturing, counting and analysis or freezing
  • Individually packed in easy to open bags

 

For preparation of:

  • Single cell suspensions of blood cells
  • Specimens for primary cell cultures and immunogens
  • Primary cells from tissue

 

Specifications

Specifications

Nylon
Polypropylene
Sterile
Conical Tubes, 50 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'