Corning™ Cell Culture Phosphate Buffered Saline (1X)
With low endotoxin content. Without calcium and magnesium.
Supplier: Corning™ 21040CM
Description
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate. PBS helps to maintain a constant pH and is isotonic and non-toxic to cells. This formulation does not contain calcium and magnesium.Specifications
2° to 30°C | |
6 x 1000 mL | |
7.3 to 7.5 |
1 X | |
Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L | |
Liquid |