missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Cell Culture Phosphate Buffered Saline (1X)
With low endotoxin content. Without calcium and magnesium.
$179.21 - $296.67
Specifications
| Concentration | 1 X |
|---|---|
| pH | 7.3 to 7.5 |
| Physical Form | Liquid |
| Formulation | Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L |
Description
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate. PBS helps to maintain a constant pH and is isotonic and non-toxic to cells. This formulation does not contain calcium and magnesium.Specifications
| 1 X | |
| 7.3 to 7.5 | |
| Liquid | |
| Potassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L |