Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine
Guaranteed low endotoxin levels ≤0.025EU/mL
Supplier:  Corning™ 10040CM
 View more versions of this product
Catalog No. MT10040CM
Explore available promotions
Description
Description
  • Ready-to-use media for mammalian in vitro cell culture is supplied in polyethylene teraphthalate (PET) plastic bottles
  • Sonically welded cap liners eliminate toxic resins or glues that could leach into media
  • Shipped at ambient temperature
Specifications
Specifications
2° to 8°C
RPMI 1640
None
6 × 1 L
Liquid
Glutamine
7.2 to 7.4