Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.
Supplier:  Corning Incorporated 25055CM
 View more versions of this product
Catalog No. MT25055CM
Explore available promotions
Alert:
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Description
Description
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Specifications
Specifications
Bottle
6 x 1 L