Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.
$134.86 - $1654.70
Specifications
Packaging Bottle
Quantity 6 x 100 mL
Alert:
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MT25055CI
View Documents
Corning Incorporated
25055CI
6 x 100 mL Bottle
Case of 6 Each for $154.80
MT25055CV
View Documents
Corning Incorporated
25055CV
6 x 500 mL Bottle
Case of 6 Each for $136.40
MT25055CVC
View Documents
Corning Incorporated
25055CVC
6 x 500 mL Bottle with septum cap
Pack of 6 Each for $134.86
MT25055CM
View Documents
Corning Incorporated
25055CM
6 x 1 L Bottle
Case of 6 Each for $283.26
MT25055LB
View Documents
Corning Incorporated
25055LB
20 L Bioprocess Container
Each for $449.60
MT25055LG
View Documents
Corning Incorporated
25055LG
1 x 100 L Bioprocess Container
Each for $1,146.93
MT25055LH
View Documents
Corning Incorporated
25055LH
1 x 200 L Bottle
Each for $1,654.70
Description
Description
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Order Info
Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.
Specifications
Specifications
Bottle
6 x 100 mL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'