Corning™ Frosted Microscope Slides
Made from water-white glass to maximize clarity. Easy to write on.
Supplier: Corning™ 294975X25
Description
These Corning™ microscope slides are made from water-white glass to maximize clarity and are frosted at one end, both sides. Slides are 0.9 to 1.10 mm thick. Corning™ frosted slides are highly legible and easy to write on because they are sandblasted instead of etched. To minimize waste, they are inspected for chips and rough edges before being packaged. Products are packaged in 1/2 gross boxes (approx. 72 slides each). 10 gross = 20 boxes (1440 slides total)Specifications
Soda Lime Glass | |
One End, Both Sides | |
72 | |
25 mm |
1440 | |
0.9 to 1.1 mm | |
75 mm |