Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable
Designed for universal fit with all common brands of pipet tips
Supplier:  Corning™ 4071
 View more versions of this product
Catalog No. 07-764-701
Explore available promotions
Description
Description
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
Warranty
3 year
Specifications
Specifications
±4 to ±0.5%
10μL Tip
10 μL Tip
Lambda™ Plus
0.5 to 10 μL
Key is included with each pipette for easy in-house calibration
3 Years
ISO8655
Autoclavable
1
Single-Channel Pipettor
10μL Tip
Yes
Videos
SDS
Documents
Documents
Accessories