Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable
Designed for universal fit with all common brands of pipet tips
Supplier: Corning™ 4073
Description
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
- Factory calibrated to strict ISO 8655 standards-3-year warranty
- Fully autoclavable and UV resistant
- Designed for universal fit with all common brands of pipet tips
- Adjustable tip ejector
Warranty
3 year
Specifications
±1.6 to ±0.8% | |
200μL Tip | |
200 μL Tip | |
Lambda™ Plus | |
10 to 100 μL | |
Key is included with each pipette for easy in-house calibration | |
3 Years |
ISO8655 | |
Autoclavable | |
1 | |
Single-Channel Pipettor | |
200μL Tip | |
Yes |