Corning™ Costar™ Low Binding Plastic Microcentrifuge Tubes
Feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in a better sample recovery. Corning™ Costar™ Low Binding Plastic Microcentrifuge Tubes™ withstand forces up to 17,000 x g.
Supplier:  Corning™ 3207
 View more versions of this product
Catalog No. 07-200-184
Explore available promotions
Description
Description
Feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in a better sample recovery. Corning™ Costar™ Low Binding Plastic Microcentrifuge Tubes™ withstand forces up to 17,000 x g.
  • Supplied nonsterile and are autoclavable.
  • External graduations and frosted writing spot for easy sample identification.
  • Positive seal design allows repeated opening and closing.
  • Flat cap surface for convenient labeling.
  • Withstands a maximum RCF of 17,000 x g.
  • Costar low binding microcentrifuge tubes feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in better sample recovery.
  • RNase-/DNase-free.
Specifications
Specifications
Snap Cap
Natural
Autoclavable
Natural
250/Pk.
Yes
Conical
Microcentrifuge Tube
RNase, DNase-free
Polypropylene
1.7 mL
Polypropylene
Non-sterile
Snap Cap
17,000 x g
Product Suggestions
Product Suggestions