Corning™ 384-Well Clear Polystyrene Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
Supplier:  Corning™ C3680
 View more versions of this product
Catalog No. 07-201-157
Explore available promotions
Description
Description
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • Specifications
    Specifications
    Standard
    0.06 cm2
    Clear
    384
    Sterile
    Flat Bottom (F-well)
    100/Cs.
    100/Cs.
    With Lid
    16 x 24
    Polystyrene
    20 to 80 μL
    Non-treated
    Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    20/Pk.