missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 384-Well Clear Polystyrene Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
$2337.63 - $2768.78
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Color | Clear |
Material | Polystyrene |
No. of Wells | 384 |
Catalog Number | Mfr. No. | Surface Treatment | Lid | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Treatment | Lid | Price | Quantity | ||||
07-200-643
|
Corning™
C3700 |
High Binding | Without Lid |
Case of 100 Each for $2,768.78
|
|
||||
07-200-645
|
Corning™
C3702 |
Non-treated | Without Lid |
Case of 100 Each for $2,337.63
|
|
||||
07-201-157
|
Corning™
C3680 |
Non-treated | With Lid |
Case of 100 Each for $2,649.65
|
|
||||
Description
Specifications
Standard | |
Clear | |
384 | |
Flat Bottom (F-well) | |
100/Cs. |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
100/Cs. |