Corning™ PYREX™ VISTA™ Round-Bottom Boiling Flasks
Designed for great shock resistance.
$98.95 - $98.95
Specifications
Material | Borosilicate Glass |
---|---|
Neck Style | Short |
Standard Taper Joint | 24/40 |
Disposable | Reusable |
Bottom Shape | Round |
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
S76105D
|
Corning™
703201LEM |
1000 mL | 24/40 |
Each for $98.95
Case of 2 Each for $197.90
|
|
||||
Description
- Full length standard taper outer joints
- Short necks
- Manufactured from No. 7740 borosilicate glass
- Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
Specifications
Borosilicate Glass | |
24/40 | |
Round |
Short | |
Reusable |