Corning™ PYREX™ VISTA™ Round-Bottom Boiling Flasks
Designed for great shock resistance.
Supplier:  Corning™ 703201LEM
 View more versions of this product
Catalog No. S76105D
Explore available promotions
Description
Description
  • Full length standard taper outer joints
  • Short necks
  • Manufactured from No. 7740 borosilicate glass
  • Wall thicknesses provide balance between thermal expansion and mechanical strength
Compliance
Designed to ASTM™ E-1403.
Specifications
Specifications
1000 mL
Short
Reusable
Borosilicate Glass
24/40
Round