missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Reusable Plastic Beakers with Handle and Spout

Translucent polypropylene beakers printed with blue graduations

Supplier:  Corning™ 1015P3L

 View more versions of this product

Catalog No. 10-210-679


Explore available promotions

Description

Description

Translucent polypropylene beakers printed with blue graduations.

Specifications

Specifications

Translucent
17 cm
Polypropylene
9.5 in.
Handle, Spout
68°F
Beaker
Reusable
DIN, ASTM Class A and B
Griffin
6.6 in.
3000 mL
For High Temperature and Chemical Resistance
24.3 cm
50 mL
20°C
Autoclavable
Yes
Blue Graduated Scale
Videos
SDS
Documents

Documents

Promotions

Promotions