missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Reusable Plastic Beakers with Handle and Spout

Translucent polypropylene beakers printed with blue graduations

$255.23 - $362.32

Specifications

Color Translucent
Material Polypropylene
For Use With (Application) For High Temperature and Chemical Resistance
Includes Handle, Spout
Temperature (English) Max. Operating 68°F
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
10-210-675
View Documents
Corning™
1015P250
250 mL
Case of 12 Each for $341.27
 
10-210-676
View Documents
Corning™
1015P500
500 mL
Case of 12 Each for $297.70
 
10-210-677
View Documents
Corning™
1015P1L
1 L
Case of 12 Each for $362.32
 
10-210-678
View Documents
Corning™
1015P2L
2000 mL
Case of 8 Each for $298.60
 
10-210-679
View Documents
Corning™
1015P3L
3000 mL
Case of 6 Each for $255.23
 
Description

Description

Translucent polypropylene beakers printed with blue graduations.

Specifications

Specifications

Translucent
For High Temperature and Chemical Resistance
68°F
Beaker
Reusable
DIN, ASTM Class A and B
Griffin
Polypropylene
Handle, Spout
20°C
Autoclavable
Yes
Blue Graduated Scale
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'