Corning™ Universal Fit Pipet Tips: Bulk Packed
![Edge](/content/dam/fishersci/glyphs/edge.png)
Designed to assure a reliable fit with major brand pipetters
Supplier: Corning™ C4845
Description
Designed to assure a reliable fit with major brand pipetters
- Beveled orifice helps ensure accurate fluid delivery
- 1–200μL tips are graduated at 10, 50 and 100μL
- 100–1000μL tips are graduated at 250, 500, and 1000μL
- Choice of sterile or nonsterile
- Bulk packed tips are nonsterile and very economical
Specifications
1 to 200 μL | |
Yellow | |
Round | |
Yes |
Non-sterile | |
Bulk | |
Universal Fit | |
Polypropylene |