missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Universal Fit Pipet Tips: Bulk Packed
Designed to assure a reliable fit with major brand pipetters
$66.65 - $707.96
Specifications
Sterility | Non-sterile |
---|---|
Format | Bulk |
For Use With (Equipment) | Universal Fit |
Graduated | Yes |
Material | Polypropylene |
Catalog Number | Mfr. No. | Format | Volume (Metric) | Color | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Volume (Metric) | Color | Price | Quantity | |||
07-200-293
|
Corning™
4844 |
Bulk | 1 to 200 μL | Natural |
Case of 10000 Each for $599.07
|
|
|||
07-200-294
![]() |
Corning™
C4845 |
Bulk | 1 to 200 μL | Yellow |
Case of 10000 Each for $608.21
|
![]() |
|||
07-200-295
|
Corning™
C4846 |
Bulk | 100 to 1000 μL | Blue |
Case of 10000 Each for $707.96
|
|
|||
07-200-296
|
Corning™
C4862 |
Bulk | 1 to 200 μL | Natural |
Case of 1000 Each for $67.83
|
|
|||
07-200-297
|
Corning™
C4866 |
Bulk | 1 to 200 μL | Yellow |
Case of 1000 Each for $66.65
|
|
|||
07-200-298
|
Corning™
C4868 |
Bulk | 100 to 1000 μL | Blue |
Case of 1000 Each for $82.97
|
|
|||
Description
Designed to assure a reliable fit with major brand pipetters
- Beveled orifice helps ensure accurate fluid delivery
- 1–200μL tips are graduated at 10, 50 and 100μL
- 100–1000μL tips are graduated at 250, 500, and 1000μL
- Choice of sterile or nonsterile
- Bulk packed tips are nonsterile and very economical
Specifications
Non-sterile | |
Universal Fit | |
Polypropylene |
Bulk | |
Yes |