Corning™ Universal Fit Pipet Tips: Bulk Packed
Designed to assure a reliable fit with major brand pipetters
Supplier:  Corning™ C4868
 View more versions of this product
Catalog No. 07-200-298
Description
Description
Designed to assure a reliable fit with major brand pipetters
  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical
Specifications
Specifications
100 to 1000 μL
Blue
Round
Yes
Non-sterile
Bulk
Universal Fit
Polypropylene