missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts

Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.

Supplier:  Corning™ C3415

 View more versions of this product

Catalog No. 07-200-148


Explore available promotions

Description

Description

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
Specifications

Specifications

12 inserts/24 well plate
Polycarbonate
3 μm
Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
4 plates; 12 inserts/plate
Clear
Sterile
0.33 cm2
0.6 mL
Pre-loaded Microporous Insert in Tissue Culture Plate
6.5 mm
TC-Treated
TC-Treated
24
Videos
SDS
Documents

Documents

Promotions

Promotions