missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts

Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.

$636.22 - $1185.08

Specifications

Material Polycarbonate
Type Pre-loaded Microporous Insert in Tissue Culture Plate
For Use With (Application) Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
Surface Coating TC-Treated
Color Clear
View More Specs

Products 9
Catalog Number Mfr. No. Diameter (Metric) Outer Pore Size Culture Area Description Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Pore Size Culture Area Description Price Quantity  
07-200-165
View Documents
Corning™
3412
24.5 mm 0.4 μm 4.7 cm2 6 inserts in 6 well plate; 4 plates/Cs.
Case of 24 Each for $636.22
 
07-200-156
View Documents
Corning™
C3401
12 mm 0.4 μm 1.12 cm2 12 inserts/12 well plate
Case of 48 Each for $1,060.93
 
07-200-157
View Documents
Corning™
3402
12 mm 3 μm 1.12 cm2 12 inserts/12 well plate
Case of 48 Each for $1,185.08
 
07-200-169
View Documents
Corning™
3428
24.5 mm 8 μm 4.7 cm2 6 inserts/6 well plate
Case of 24 Each for $668.44
 
07-200-148
View Documents
Corning™
C3415
6.5 mm 3 μm 0.33 cm2 12 inserts/24 well plate
Case of 48 Each for $1,134.17
 
07-200-149
View Documents
Corning™
3421
6.5 mm 5 μm 0.33 cm2 12 inserts/24 well plate
Case of 48 Each for $1,097.81
 
07-200-147
View Documents
Corning™
3413
6.5 mm 0.4 μm 0.33 cm2 12 inserts/24 well plate
Case of 48 Each for $1,092.91
 
07-200-150
View Documents
Corning™
C3422
6.5 mm 8 μm 0.33 cm2 12 inserts in a 24 well plate; 4 plates per case
Case of 48 Each for $1,124.35
 
07-200-166
View Documents
Corning™
C3414
24.5 mm 3 μm 4.7 cm2 6 inserts/6 well plate
Case of 24 Each for $655.08
 
Description

Description

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
Specifications

Specifications

Polycarbonate
Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
Clear
Sterile
Pre-loaded Microporous Insert in Tissue Culture Plate
TC-Treated
TC-Treated
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'