Thermo Scientific™ Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Supplier:  Thermo Scientific™ GPSSL02
 View more versions of this product
Catalog No. GPSSL02
Explore available promotions
Description
Description
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Features:
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
  • Enhanced hinged design supports open-lid configuration and easy lid removal
  • Accommodate taller labware with extended height
  • Polycarbonate and stainless steel models available
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Stainless Steel Gable Cover
Precision 2L General Purpose Water Bath
Stainless Steel
2 L
Gable Cover