StatLab™ CoverSafe Mounting Medium
A fast-drying, low-hazard, optically clear, permanent mounting medium.
Supplier: StatLab™ MMC0226
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Description
- CoverSafe contains no xylene, no toluene, no dioxane, and no benzene
- Pleasant citrus scent, making it an ideal low-hazard permanent mounting medium
- For use in histology and cytology
Specifications
Mounting Medium | |
Mounting Solution | |
100 mL |