Covidien Chemosafety™ Containers
For disposal of chemotherapy waste
Supplier:  Covidien 8985
 View more versions of this product
Catalog No. 14-830-137
Explore available promotions
Description
Description
  • Yellow color makes container highly identifiable as chemotherapy waste
  • Accommodates assorted sizes of waste materials and sharps, including syringes, ampules, and prep materials
  • Sealing gasketed lid provides leak-resistant containment of fluid
  • Container locks to make it secure for final disposal
  • Molded handles aid in secure transport
  • Nestable containers save valuable storage space
Specifications
Specifications
30.2 L
8 gal.
For disposal of trace chemotherapy waste
Sharps Disposal Container
Hinged Top
Yellow
Polypropylene