Covidien Defender™ Safety Needle Holders
Total needle protection allows for safe, efficient and easy use
Supplier:  Covidien 8881225110
 View more versions of this product
Catalog No. 22257420
Explore available promotions
Description
Description
Single-use devices eliminate needle removal and blood tube holder reuse, and help prevent needle-stick injury
  • One-handed safety activation
  • Front and back end needle protection
  • Needle retracts into blood tube holder to reduce sharps disposal space
  • May be used with 1 in. or 1.5 in. standard blood collection needles
  • Latex free
Compliance
Meets FDA Quality System Requirements (QSR) per 21 Code of Federal Regulations (CFR) Part 820. Complies with Occupational Safety and Health Administration (OSHA) Standards.
Specifications
Specifications
Single-use
Needle Holder
Blue
1 in.
Provide front and back end needle protection
2.5 cm