Covidien RCRA Hazardous Waste Containers
Effectively segregate hazardous pharmaceutical waste in accordance with the Resource Conservation and Recovery Act (RCRA)
Supplier: Covidien 8608RC
Description
- Black container with white lid makes it easy to distinguish from sharps, chemo, and other disposal containers
- Leak-resistant gasket and absorbent pad help contain liquid contents
Compliance
RCRA Hazardous Waste Label meets EPA satellite accumulation labeling requirements. Meets DOT requirements for transport without secondary packaging.
Specifications
RCRA, EPA, DOT | |
8 gal. | |
Gasket, absorbent pad, and extra labels, sliding lid | |
Waste Container |
Slide Top | |
Black | |
Segregate hazardous pharmaceutical waste under the Resource Conservation and Recovery Act |