missing translation for 'onlineSavingsMsg'
Learn More

Covidien RCRA Hazardous Waste Containers

Effectively segregate hazardous pharmaceutical waste in accordance with the Resource Conservation and Recovery Act (RCRA)

$43.09 - $190.22

Specifications

Certifications/Compliance RCRA, EPA, DOT
Color Black
For Use With (Application) Segregate hazardous pharmaceutical waste under the Resource Conservation and Recovery Act
Product Type Waste Container
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (English) Closure Type Price Quantity  
Catalog Number Mfr. No. Capacity (English) Closure Type Price Quantity  
22-031-376
View Documents
Covidien
8602RC
2 gal. Hinged Lid with Snap Cap
Each for $43.09
Case of 20 Each for $861.80
Only null left
 
22-031-377
View Documents
Covidien
8608RC
8 gal. Slide Top
Each for $125.45
Case of 10 Each for $1,254.50
Only null left
 
22-031-379
View Documents
Covidien
8618RC
18 gal. Slide Top
Each for $190.22
Case of 5 Each for $951.10
Only null left
 
22-031-378
View Documents
Covidien
8612RC
12 gal. Slide Top
Each for $101.09
Case of 10 Each for $1,010.90
Only null left
 
Description

Description

  • Black container with white lid makes it easy to distinguish from sharps, chemo, and other disposal containers
  • Leak-resistant gasket and absorbent pad help contain liquid contents

Compliance

RCRA Hazardous Waste Label meets EPA satellite accumulation labeling requirements. Meets DOT requirements for transport without secondary packaging.

Specifications

Specifications

RCRA, EPA, DOT
Black
Segregate hazardous pharmaceutical waste under the Resource Conservation and Recovery Act
Waste Container
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'