Covidien RCRA Hazardous Waste Containers
Effectively segregate hazardous pharmaceutical waste in accordance with the Resource Conservation and Recovery Act (RCRA)
Supplier:  Covidien 8612RC
 View more versions of this product
Catalog No. 22-031-378
Explore available promotions
Description
Description
  • Black container with white lid makes it easy to distinguish from sharps, chemo, and other disposal containers
  • Leak-resistant gasket and absorbent pad help contain liquid contents
Compliance
RCRA Hazardous Waste Label meets EPA satellite accumulation labeling requirements. Meets DOT requirements for transport without secondary packaging.
Specifications
Specifications
RCRA, EPA, DOT
12 gal.
Leak-resistant gasket, absorbent pad, and extra labels
Waste Container
Slide Top
Black
Segregate hazardous pharmaceutical waste under the Resource Conservation and Recovery Act