Innovatek C-Reactive Protein (CPR) Latex Test Kits
Innovatek C-Reactive Protein (CPR) Latex Test Kits offer results in less than 3 minutes
Supplier:  Innovatek 419110103
 View more versions of this product
Catalog No. 29-801-066
Explore available promotions
Description
Description
  • Fast 2 to 3 minute rotation
  • Complete kit includes all reagents and controls
  • Disposable test cards
  • 6 to 12 months of shelf life
  • Sensitive and specific
Specifications
Specifications
C-Reactive Protein (CRP)
Single-use
100 Tests/Kit
≈3 min.