missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Creatinine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 228940250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Creatinine | |
255.0°C | |
290°C | |
Authentic | |
Glass bottle | |
25 g | |
15,2557 | |
(5% in water) Clear colorless to slightly yellow | |
CN1CC(=O)N=C1N | |
113.12 | |
CHEBI:16737 | |
98% |
60-27-5 | |
White | |
0.5% (105°C) max. | |
98% | |
C4H7N3O | |
24,245 | |
creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu | |
DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
2-amino-3-methyl-4H-imidazol-5-one | |
588 | |
113.12 | |
Crystalline Powder |
Chemical Identifiers
60-27-5 | |
113.12 | |
creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu | |
CHEBI:16737 | |
CN1CC(=O)N=C1N |
C4H7N3O | |
DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
588 | |
2-amino-3-methyl-4H-imidazol-5-one |
Safety and Handling
EINECSNumber : 200-466-7
RUO – Research Use Only