missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatinine, 98%
CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
$101.84 - $678.71
Chemical Identifiers
| CAS | 60-27-5 |
|---|---|
| Molecular Formula | C4H7N3O |
| Molecular Weight (g/mol) | 113.12 |
| InChI Key | DDRJAANPRJIHGJ-UHFFFAOYSA-N |
| Synonym | creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu |
| PubChem CID | 588 |
| ChEBI | CHEBI:16737 |
| IUPAC Name | 2-amino-3-methyl-4H-imidazol-5-one |
| SMILES | CN1CC(=O)N=C1N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC228940250
|
Thermo Scientific Chemicals
228940250 |
25 g | Glass bottle |
Each for $101.84
|
|
||||
|
AC228940500
|
Thermo Scientific Chemicals
228940500 |
50 g | Glass bottle |
Each for $171.14
|
|
||||
|
AC228942500
|
Thermo Scientific Chemicals
228942500 |
250 g | Glass bottle |
Each for $678.71
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 60-27-5 | |
| 113.12 | |
| creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu | |
| CHEBI:16737 | |
| CN1CC(=O)N=C1N |
| C4H7N3O | |
| DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
| 588 | |
| 2-amino-3-methyl-4H-imidazol-5-one |
Specifications
| 60-27-5 | |
| White | |
| 0.5% (105°C) max. | |
| 98% | |
| C4H7N3O | |
| 24,245 | |
| creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu | |
| DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
| 2-amino-3-methyl-4H-imidazol-5-one | |
| 588 | |
| 113.12 | |
| Crystalline Powder |
| 255.0°C | |
| 290°C | |
| Authentic | |
| Glass bottle | |
| 25 g | |
| 15,2557 | |
| (5% in water) Clear colorless to slightly yellow | |
| CN1CC(=O)N=C1N | |
| 113.12 | |
| CHEBI:16737 | |
| 98% | |
| Creatinine |
Safety and Handling
EINECSNumber : 200-466-7
RUO – Research Use Only