missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Creatinine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 228942500

 View more versions of this product

Catalog No. AC228942500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Creatinine
255.0°C
290°C
Authentic
Glass bottle
250 g
15,2557
(5% in water) Clear colorless to slightly yellow
CN1CC(=O)N=C1N
113.12
CHEBI:16737
98%
60-27-5
White
0.5% (105°C) max.
98%
C4H7N3O
24,245
creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
DDRJAANPRJIHGJ-UHFFFAOYSA-N
2-amino-3-methyl-4H-imidazol-5-one
588
113.12
Crystalline Powder

Chemical Identifiers

60-27-5
113.12
creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
CHEBI:16737
CN1CC(=O)N=C1N
C4H7N3O
DDRJAANPRJIHGJ-UHFFFAOYSA-N
588
2-amino-3-methyl-4H-imidazol-5-one
Safety and Handling

Safety and Handling

EINECSNumber : 200-466-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only